Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-methyl-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D0583500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0583500G 500 g
D058325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0583500G Supplier TCI America Supplier No. D0583500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 542-02-9
Molecular Formula C4H7N5
Molecular Weight (g/mol) 125.135
MDL Number MFCD00023192
InChI Key NJYZCEFQAIUHSD-UHFFFAOYSA-N
Synonym acetoguanamine, 2,4-diamino-6-methyl-1,3,5-triazine, 1,3,5-triazine-2,4-diamine, 6-methyl, 2-methyl-4,6-diamino-1,3,5-triazine, 2,4-diamino-6-methyl-s-triazine, 6-methyl-1,3,5-triazine-2,4-diyldiamine, s-triazine, 2,4-diamino-6-methyl, acetoguanamin, ccris 8909, dsstox_cid_9186
PubChem CID 10949
ChEBI CHEBI:72475
IUPAC Name 6-methyl-1,3,5-triazine-2,4-diamine
SMILES CC1=NC(=NC(=N1)N)N

Specifications

Color White
Quantity 500 g
Formula Weight 125.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-methyl-1,3,5-triazine
EINECSNumber (5)-1026
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.