accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-16-5

IUPAC Name: bis(aniline); sulfuric acid
Synonyms: bis(aniline); sulfuric acid
Molecular Weight (g/mol): 284.33
Molecular Formula: C12H16N2O4S
InChi Key: FUKMEFZGEMVGLD-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC=CC=C1.NC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)