Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline Sulfate 99%, Thermo Scientific™

Catalog No. AC401205000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC401205000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC401205000 Supplier Thermo Scientific Chemicals Supplier No. 401205000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-16-5
Molecular Formula C12H14N2·H2SO4
MDL Number MFCD00013110

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 500 g
Merck Index 15, 652
Formula Weight 284.33
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Aniline sulfate, 99%
Hazard Category Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction. Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure. Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed. Causes serious eye damage. Suspected of causing genetic defects. Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid release to the environment.
EINECSNumber 208-805-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.