CAS RN 542-44-9
IUPAC Name:
2,3-dihydroxypropyl hexadecanoate
Synonyms:
glyceryl palmitate
Molecular Weight (g/mol):
330.51
Molecular Formula:
C19H38O4
InChi Key:
QHZLMUACJMDIAE-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)OCC(O)CO
1
–
1
of
1
results
1
Monopalmitin 95.0+%, TCI America™
CAS: 542-44-9 Molecular Formula: C19H38O4 Molecular Weight (g/mol): 330.51 MDL Number: MFCD00042734 InChI Key: QHZLMUACJMDIAE-UHFFFAOYNA-N Synonym: Glycerol alpha-Monopalmitate PubChem CID: 14900 ChEBI: CHEBI:69081 IUPAC Name: 2,3-dihydroxypropyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)CO