Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monopalmitin 95.0+%, TCI America™
SDP

Catalog No. G008325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G008325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G008325G Supplier TCI America Supplier No. G008325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 542-44-9
Molecular Formula C19H38O4
Molecular Weight (g/mol) 330.51
MDL Number MFCD00042734
InChI Key QHZLMUACJMDIAE-UHFFFAOYNA-N
Synonym Glycerol alpha-Monopalmitate
PubChem CID 14900
ChEBI CHEBI:69081
IUPAC Name 2,3-dihydroxypropyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)CO

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 25 g
Formula Weight 330.51
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monopalmitin
EINECSNumber (2)-0669
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.