accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-58-5

IUPAC Name: 2-chloroethyl acetate
Synonyms: ethanol, 2-chloro-, acetate
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: VIRWKAJWTKAIMA-UHFFFAOYSA-N
SMILES: CC(=O)OCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results