Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Acetate 99.0+%, TCI America™
SDP

Catalog No. A002725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A002725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A002725ML Supplier TCI America Supplier No. A002725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 542-58-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00018963
InChI Key VIRWKAJWTKAIMA-UHFFFAOYSA-N
Synonym Acetic Acid 2-Chloroethyl Ester
PubChem CID 10959
IUPAC Name 2-chloroethyl acetate
SMILES CC(=O)OCCCl

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 2929
Formula Weight 122.55
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Acetate
RTECSNumber KK1010000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.