accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5421-04-5

IUPAC Name: 3-methylbutyl 2-hydroxy-2-phenylacetate
Synonyms: 3-methylbutyl 2-hydroxy-2-phenylacetate
Molecular Weight (g/mol): 222.28
Molecular Formula: C13H18O3
InChi Key: KQQXUARABJGCMS-UHFFFAOYNA-N
SMILES: CC(C)CCOC(=O)C(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results