Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl DL-Mandelate 93.0+%, TCI America™
SDP

Catalog No. M071625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M071625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M071625G Supplier TCI America Supplier No. M071625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5421-04-5
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.28
MDL Number MFCD00021858
InChI Key KQQXUARABJGCMS-UHFFFAOYNA-N
Synonym DL-Mandelic Acid Isoamyl Ester, Isopentyl DL-Mandelate, DL-Mandelic Acid Isopentyl Ester
PubChem CID 95396
IUPAC Name 3-methylbutyl 2-hydroxy-2-phenylacetate
SMILES CC(C)CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 169°C
Quantity 25 g
Formula Weight 222.28
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl DL-Mandelate
RTECSNumber CY1633900
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.