accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CNH2NH2NNCH3NH2H2NHClHCl(E/Z)(E/Z)

CAS RN 5421-66-9

IUPAC Name: 4-(2-{3-[2-(2,4-diamino-5-methylphenyl)diazen-1-yl]-4-methylphenyl}diazen-1-yl)-6-methylbenzene-1,3-diamine dihydrochloride
Synonyms: bismarck brown Y dihydrochloride
Molecular Weight (g/mol): 461.4
Molecular Formula: C21H26Cl2N8
InChi Key: WLKAMFOFXYCYDK-UHFFFAOYSA-N
SMILES: Cl.Cl.CC1=CC(N=NC2=CC(N=NC3=C(N)C=C(N)C(C)=C3)=C(C)C=C2)=C(N)C=C1N
Color 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)