Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismarck Brown R; MP Biomedicals

Catalog No. ICN15485880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15485880 100 g
ICN15485850 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15485880 Supplier MP Biomedicals, Inc Supplier No. 0215485880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5421-66-9
Molecular Formula C21H26Cl2N8
Molecular Weight (g/mol) 461.395
InChI Key WLKAMFOFXYCYDK-UHFFFAOYSA-N
Synonym C.I. 21010, Basic Brown 4
PubChem CID 79459
IUPAC Name 4-[[3-[(2,4-diamino-5-methylphenyl)diazenyl]-4-methylphenyl]diazenyl]-6-methylbenzene-1,3-diamine;dihydrochloride
SMILES CC1=C(C=C(C=C1)N=NC2=C(C=C(C(=C2)C)N)N)N=NC3=C(C=C(C(=C3)C)N)N.Cl.Cl

Specifications

Assay Percent Range ∽ 60%
Color Black/Blue to Blue
Quantity 100 g
Formula Weight 461.4
Grade Not Available
Chemical Name or Material Bismarck Brown R
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.