accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5422-34-4

IUPAC Name: 2-hydroxy-N-(2-hydroxyethyl)propanamide
Synonyms: lactamide mea N-(2-hydroxyethyl)lactamide
Molecular Weight (g/mol): 133.15
Molecular Formula: C5H11NO3
InChi Key: RZCHTMXTKQHYDT-UHFFFAOYNA-N
SMILES: CC(O)C(=O)NCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results