Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Hydroxyethyl)lactamide 98.0+%, TCI America™
SDP

Catalog No. H037825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H037825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H037825G Supplier TCI America Supplier No. H037825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5422-34-4
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.147
MDL Number MFCD00037805
InChI Key RZCHTMXTKQHYDT-UHFFFAOYSA-N
Synonym N-(2-Hydroxyethyl)-2-hydroxypropionamide
PubChem CID 95457
IUPAC Name 2-hydroxy-N-(2-hydroxyethyl)propanamide
SMILES CC(C(=O)NCCO)O

Specifications

Quantity 25 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)lactamide
EINECSNumber (2)-1378
RTECSNumber OD2400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.