accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CSOONNN

CAS RN 54230-59-0

IUPAC Name: 1-(2,4,6-trimethylbenzenesulfonyl)-1H-1,2,4-triazole
Synonyms: 1-(2,4,6-trimethylbenzenesulfonyl)-1,2,4-triazole
Molecular Weight (g/mol): 251.3
Molecular Formula: C11H13N3O2S
InChi Key: XNKYPZJMRHXJJQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results