Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Mesitylsulfonyl)-1H-1,2,4-triazole 99%, Thermo Scientific™

Catalog No. AC210060010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC210060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC210060010 Supplier Thermo Scientific Chemicals Supplier No. 210060010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54230-59-0
Molecular Formula C11H13N3O2S
Molecular Weight (g/mol) 251.304
InChI Key XNKYPZJMRHXJJQ-UHFFFAOYSA-N
Synonym 1-mesitylsulfonyl-1h-1,2,4-triazole, 1-2-mesitylenesulfonyl-1h-1,2,4-triazole, 1-mesitylen-2-sulfonyl-1h-1,2,4-triazole, 1-2-mesitylenesulfonyl-1,2,4-triazole, mst, 1-2-mesitylenesulfonyl-1h-1,2,4-triazole, n-mesitylenesulfonyl-1,2,4-triazole, 1-2-mesitylsulfonyl-1h-1,2,4-triazole, 1-2,4,6-trimethylphenyl sulfonyl-1,2,4-triazole, 1-2,4,6-trimethylbenzenesulfonyl-1,2,4-triazole, mesityl 1,2,4-triazole-1-sulphonate
PubChem CID 521491
IUPAC Name 1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=NC=N2)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 130°C to 134°C
Color White
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 251.31
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-(2-Mesitylsulfonyl)-1H-1, 2, 4-triazole, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.