accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54230-60-3

H3CCH3CH3CH3H3CCH3SOONNN

CAS RN 54230-60-3

IUPAC Name: 1-[2,4,6-tris(propan-2-yl)benzenesulfonyl]-1H-1,2,4-triazole
Synonyms: 1-(2,4,6-triisopropylbenzenesulfonyl)-1,2,4-triazole
Molecular Weight (g/mol): 335.47
Molecular Formula: C17H25N3O2S
InChi Key: BKJMMUUBRUWPPQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)N1C=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results