Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4,6-Triisopropylbenzenesulfonyl)-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. T29515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29515G 5 g
T29511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29515G Supplier TCI America Supplier No. T29515G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 54230-60-3
Molecular Formula C17H25N3O2S
Molecular Weight (g/mol) 335.47
MDL Number MFCD00014089
InChI Key BKJMMUUBRUWPPQ-UHFFFAOYSA-N
Synonym 1-2,4,6-tris isopropyl phenyl sulphonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylphenyl sulfonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylbenzenesulfonyl-1,2,4-triazole, tpst, 2,4,6-triisopropylbenzenesulfonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylphenyl-sulfonyl-1h-1,2,4-triazole, 1-2,4,6-triisopropylphenylsulfonyl-1,2,4-triazole, 1-2,4,6-tri propan-2-yl phenyl sulfonyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-2,4,6-tris 1-methylethyl phenyl sulfonyl
PubChem CID 688654
IUPAC Name 1-[2,4,6-tris(propan-2-yl)benzenesulfonyl]-1H-1,2,4-triazole
SMILES CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)N1C=NC=N1

Specifications

Melting Point 115°C
Color White
Quantity 5 g
Formula Weight 335.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,4,6-Triisopropylbenzenesulfonyl)-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.