accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3HNOHNH2N

CAS RN 54239-37-1

IUPAC Name: 2-amino-5-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzonitrile
Synonyms: cimaterol cimaterolum cimatrol
Molecular Weight (g/mol): 219.29
Molecular Formula: C12H17N3O
InChi Key: BUXRLJCGHZZYNE-UHFFFAOYNA-N
SMILES: CC(C)NCC(O)C1=CC=C(N)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results