accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBr

CAS RN 54247-27-7

IUPAC Name: {[(6-bromohexyl)oxy]methyl}benzene
Synonyms: {[(6-bromohexyl)oxy]methyl}benzene
Molecular Weight (g/mol): 271.2
Molecular Formula: C13H19BrO
InChi Key: UHDZRWPKYMHVNV-UHFFFAOYSA-N
SMILES: BrCCCCCCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results