accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH

CAS RN 54256-45-0

IUPAC Name: 1-octylpiperazine
Synonyms: 1-octylpiperazine
Molecular Weight (g/mol): 198.35
Molecular Formula: C12H26N2
InChi Key: RRZDZQLIIMBIDL-UHFFFAOYSA-N
SMILES: CCCCCCCCN1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results