CAS RN 5428-41-1
IUPAC Name:
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
Synonyms:
5-cyanouracil
Molecular Weight (g/mol):
137.1
Molecular Formula:
C5H3N3O2
InChi Key:
HAUXRJCZDHHADG-UHFFFAOYSA-N
SMILES:
O=C1NC=C(C#N)C(=O)N1
1
–
1
of
1
results
1
Promotions Available
5-Cyanouracil, 97%
5-Cyanouracil, 97%
CAS: 5428-41-1 Molecular Formula: C5H3N3O2 Molecular Weight (g/mol): 137.10 MDL Number: MFCD00051958 InChI Key: HAUXRJCZDHHADG-UHFFFAOYSA-N Synonym: Uracil-5-carbonitrile IUPAC Name: 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile SMILES: O=C1NC=C(C#N)C(=O)N1