Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyanouracil, 97%

Catalog No. AAL0849003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0849003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0849003 Supplier Thermo Scientific Chemicals Supplier No. L0849003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5428-41-1 | C5H3N3O2 | 137.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5428-41-1
Molecular Formula C5H3N3O2
Molecular Weight (g/mol) 137.10
MDL Number MFCD00051958
InChI Key HAUXRJCZDHHADG-UHFFFAOYSA-N
Synonym Uracil-5-carbonitrile
IUPAC Name 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
SMILES O=C1NC=C(C#N)C(=O)N1

Specifications

Melting Point >300°C (decomposition)
Quantity 1 g
UN Number UN3439
Beilstein 5707
Formula Weight 137.1
Percent Purity 97%
Chemical Name or Material 5-Cyanouracil
Hazard Category H302+H332
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.