accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NClN

CAS RN 5428-89-7

IUPAC Name: 5-chloropyrimidin-2-amine
Synonyms: 5-chloropyrimidin-2-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: OQZMDDKDHRIGDY-UHFFFAOYSA-N
SMILES: NC1=NC=C(Cl)C=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)