accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHCH2OHO

CAS RN 5429-56-1

IUPAC Name: 2-acetamidoprop-2-enoic acid
Synonyms: 2-acetamidoprop-2-enoic acid
Molecular Weight (g/mol): 129.12
Molecular Formula: C5H7NO3
InChi Key: UFDFFEMHDKXMBG-UHFFFAOYSA-N
SMILES: CC(=O)NC(=C)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)