Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-acetamidoacrylic acid, ≥99%, MP Biomedicals™

Catalog No. ICN19505401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19505401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19505401 Supplier MP Biomedicals, Inc Supplier No. 0219505401
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5429-56-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
InChI Key UFDFFEMHDKXMBG-UHFFFAOYSA-N
Synonym 2-acetamidoacrylic acid, 2-propenoic acid, 2-acetylamino, acrylic acid, 2-acetamido, 2-acetamidoacrylicacid, acetyldehydroalanine, n-acetyldehydroalanine, 2-acetylamino acrylic acid, .alpha.-acetamidoacrylic acid, 2-acetylamino-2-propenoic acid, 2-acetylamino prop-2-enoic acid
PubChem CID 79482
IUPAC Name 2-acetamidoprop-2-enoic acid
SMILES CC(=O)NC(=C)C(=O)O

Specifications

Color Cream
Quantity 1 g
Formula Weight 129.1
Physical Form Powder
Chemical Name or Material 2-Acetamidoacrylic acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.