accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 543-75-9

IUPAC Name: 2,3-dihydro-1,4-dioxine
Synonyms: 1,4-dioxene
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: HIZVCIIORGCREW-UHFFFAOYSA-N
SMILES: C1COC=CO1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results