Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxene 98.0+%, TCI America™
SDP

Catalog No. D22875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22875G 5 g
D22871G 1 g
D228725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22875G Supplier TCI America Supplier No. D22875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 543-75-9
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00082537
InChI Key HIZVCIIORGCREW-UHFFFAOYSA-N
Synonym 1,4-dioxene, 1,4-dioxin, 2,3-dihydro, p-dioxin, 2,3-dihydro, 2,3-dihydro-1,4-dioxin, 1,4-dioxane-2,3-diyl, #
PubChem CID 136353
IUPAC Name 2,3-dihydro-1,4-dioxine
SMILES C1COC=CO1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
UN Number 1993
Formula Weight 86.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dioxene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.