accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 543-90-8

IUPAC Name: cadmium(2+) diacetate
Synonyms: cadmium acetate
Molecular Weight (g/mol): 230.5
Molecular Formula: C4H6CdO4
InChi Key: LHQLJMJLROMYRN-UHFFFAOYSA-L
SMILES: [Cd++].CC([O-])=O.CC([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results