Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog No. AC454002500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454002500 250 g
AC454000010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454002500 Supplier Thermo Scientific Chemicals Supplier No. 454002500
  • $183.00
    $146.65 / Each of 1

    Save $36.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 543-90-8 | C4H6CdO4 | 230.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-90-8
Molecular Formula C4H6CdO4
Molecular Weight (g/mol) 230.5
InChI Key LHQLJMJLROMYRN-UHFFFAOYSA-L
Synonym cadmium acetate, cadmium diacetate, acetic acid, cadmium salt, cadmium ethanoate, bis acetoxy cadmium, cadmium di acetate, cadmium ii acetate, unii-95kc50z1l0, ccris 7783, hsdb 280
PubChem CID 10986
IUPAC Name cadmium(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Cd+2]

Specifications

Melting Point 255.0°C to 273.0°C
Quantity 250 g
Loss on Drying 0.5% max.
Assay Percent Range 97.5% min. (Complexometry)
Merck Index 15,1619
Formula Weight 230.5
Percent Purity 98%
Grade Pure
Chemical Name or Material Cadmium acetate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause cancer.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortab
EINECSNumber 208-853-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.