accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHOOHO

CAS RN 5434-20-8

IUPAC Name: 3-aminobenzene-1,2-dicarboxylic acid
Synonyms: 3-aminobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: WGLQHUKCXBXUDV-UHFFFAOYSA-N
SMILES: NC1=CC=CC(C(O)=O)=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results