Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophthalic Acid 95.0+%, TCI America™
SDP

Catalog No. A15165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15165G 5 g
A15161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15165G Supplier TCI America Supplier No. A15165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5434-20-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
InChI Key WGLQHUKCXBXUDV-UHFFFAOYSA-N
Synonym 3-amino-1,2-benzenedicarboxylic acid, phthalic acid, 3-amino, 3-aminophthalicacid, 1,2-benzenedicarboxylic acid, 3-amino, 3-amino-phthalic acid, unii-7xv0v19zdg, 3-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, amino, 7xv0v19zdg, aminophthalic acid
PubChem CID 79490
IUPAC Name 3-aminophthalic acid
SMILES C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O

Specifications

Melting Point 183°C
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.