accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5436-21-5

IUPAC Name: 4,4-dimethoxybutan-2-one
Synonyms: 2-butanone, 4,4-dimethoxy-
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: PJCCSZUMZMCWSX-UHFFFAOYSA-N
SMILES: COC(CC(C)=O)OC
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)