accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5439-11-2

IUPAC Name: 2-bromo-N-(2-chlorophenyl)acetamide
Synonyms: 2-bromo-N-(2-chlorophenyl)acetamide
Molecular Weight (g/mol): 248.5
Molecular Formula: C8H7BrClNO
InChi Key: WIOJXHICOOQVAB-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1NC(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results