Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-(2-Chlorophenyl)-2-bromoacetamide, 95%, Thermo Scientific™

Catalog No. SPB05477DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB05477DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB05477DA Supplier Thermo Scientific Chemicals Supplier No. SPB05477DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5439-11-2
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.50
MDL Number MFCD00173815
InChI Key WIOJXHICOOQVAB-UHFFFAOYSA-N
Synonym 2-bromo-n-2-chlorophenyl acetamide, n1-2-chlorophenyl-2-bromoacetamide, n-bromoacetyl-2'-chloroacetaniline, wiojxhicooqvab-uhfffaoysa-n, 2-bromo-n-2-chloro-phenyl-acetamide, acetamide, 2-bromo-n-2-chlorophenyl
PubChem CID 225525

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Linear Formula ClC6H4NHC(O)CH2Br
Quantity 1 g
Formula Weight 248.51
Percent Purity 95%
Chemical Name or Material N1-(2-chlorophenyl)-2-bromoacetamide
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.