accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(Z)(Z)

CAS RN 544-35-4

IUPAC Name: ethyl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms: ethyl linoleate ethyl linoleate, pure pure ethyl linoleate mandenol
Molecular Weight (g/mol): 308.51
Molecular Formula: C20H36O2
InChi Key: FMMOOAYVCKXGMF-MURFETPASA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)