Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Linoleate 70.0+%, TCI America™
SDP

Catalog No. L005525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L005525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L005525ML Supplier TCI America Supplier No. L005525ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 544-35-4
Molecular Formula C20H36O2
Molecular Weight (g/mol) 308.506
MDL Number MFCD00009535
InChI Key FMMOOAYVCKXGMF-MURFETPASA-N
Synonym ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
PubChem CID 5282184
ChEBI CHEBI:31572
IUPAC Name ethyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 193°C
Quantity 25 mL
Formula Weight 308.51
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Linoleate
EINECSNumber (2)-0798
RTECSNumber RG2185000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.