accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNHSCl

CAS RN 544-47-8

IUPAC Name: hydrogen {[(4-chlorophenyl)methyl]sulfanyl}methanimidamide chloride
Synonyms: hydrogen [(4-chlorophenyl)methyl]sulfanylmethanimidamide chloride
Molecular Weight (g/mol): 237.14
Molecular Formula: C8H10Cl2N2S
InChi Key: QAVHMIYSTFZJAU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)SCC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results