Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-(4-Chlorobenzyl)isothiouronium chloride, 98%, Thermo Scientific™

Catalog No. AAB2536222
Catalog No. AAB2536222 Supplier Thermo Scientific Chemicals Supplier No. B2536222

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 544-47-8
Molecular Formula C8H10Cl2N2S
MDL Number MFCD00012584
Merck Index 14,2128
Synonym unii-ug8z59jowy, p-chlorobenzylpseudothiuronium chloride, 2-4-chlorobenzyl-2-thiopseudourea hydrochloride, 2-p-chlorobenzyl isothiouronium chloride, s-4-chlorobenzyl isothiouronium chloride, ug8z59jowy, carbamimidothioic acid, 4-chlorophenyl methyl ester, monohydrochloride, 4-chlorobenzyl imidothiocarbamate hydrochloride, s-p-chlorobenzyl-thiuronium chloride, 4-chlorobenzyl carbamimidothioate hydrochloride
InChI Key QAVHMIYSTFZJAU-UHFFFAOYSA-N
SMILES [H+].[Cl-].NC(=N)SCC1=CC=C(Cl)C=C1
IUPAC Name hydrogen {[(4-chlorophenyl)methyl]sulfanyl}methanimidamide chloride
Molecular Weight (g/mol) 237.14
PubChem CID 68342
Formula Weight 237.15
Percent Purity 98%
Chemical Name or Material S-(4-Chlorobenzyl)isothiouronium chloride
Show More Show Less
RTECSNumber UM0950000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.