CAS RN 544-71-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Conjugated (9E,11E)-Linoleic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
Conjugated (9E,11E)-linoleic acid can be obtained via hydrogenation of linoleic acid in the middle [12,13] double bond.