Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conjugated (9E,11E)-Linoleic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014558 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4558 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4558 Supplier MilliporeSigma Supelco Supplier No. 9098320MG
Add to Cart
Add to Cart

Conjugated (9E,11E)-linoleic acid can be obtained via hydrogenation of linoleic acid in the middle [12,13] double bond.

TRUSTED_SUSTAINABILITY

Specifications

CAS 544-71-8
Physical Form Neat
Linear Formula C18H32O2
MDL Number MFCD00133681
Synonym (9 E,11 E)-9,11-Octadecadienoic acid; 9 E,11 E-CLA; Isolinoleic acid; Linoleic acid (9-trans, 11-trans); Mangold′s acid
Type Analytical Standard
Quantity 20 mg
Formula Weight 280.45
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.