accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH

CAS RN 5442-00-2

IUPAC Name: 2-cyclohexylpropan-1-ol
Synonyms: cyclohexylpropanol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: IRIVQXLOJHCXIE-UHFFFAOYNA-N
SMILES: CC(CO)C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results