Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclohexyl-1-propanol 99%, Thermo Scientific™

Catalog No. AC255891000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC255891000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255891000 Supplier Thermo Scientific Chemicals Supplier No. 255891000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5442-00-2
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
InChI Key IRIVQXLOJHCXIE-UHFFFAOYSA-N
Synonym 2-cyclohexyl-1-propanol, cyclohexaneethanol, .beta.-methyl, cyclohexaneethanol, beta-methyl, 2-cyclohexylpropanol, 2-methyl-2-cyclohexylethanol
PubChem CID 79509
IUPAC Name 2-cyclohexylpropan-1-ol
SMILES CC(CO)C1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4700 to 1.4720 (20°C, 589nm)
Linear Formula C6H11CH(CH3)CH2OH
Quantity 100 mL
Formula Weight 142.24
Percent Purity ≥98.5%
Physical Form Viscous Liquid
Chemical Name or Material 2-Cyclohexyl-1-propanol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.