accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OO

CAS RN 5444-75-7

IUPAC Name: 2-ethylhexyl benzoate
Synonyms: ethylhexyl benzoate
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: UADWUILHKRXHMM-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results