Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl Benzoate 99.0+%, TCI America™
SDP

Catalog No. B197725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B197725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B197725ML Supplier TCI America Supplier No. B197725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5444-75-7
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00072620
InChI Key UADWUILHKRXHMM-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Ethylhexyl Ester
PubChem CID 94310
IUPAC Name 2-ethylhexyl benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 mL
Formula Weight 234.34
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Benzoate
EINECSNumber (3)-1356&(7)-0705
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.