accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 544436-47-7

IUPAC Name: 2,7-dibromo-9-dodecyl-9H-carbazole
Synonyms: 2,7-dibromo-9-dodecylcarbazole
Molecular Weight (g/mol): 493.33
Molecular Formula: C24H31Br2N
InChi Key: NBJGUMLGJPJNLO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results