Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9-dodecylcarbazole 98.0+%, TCI America™
SDP

Catalog No. D4661200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4661200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4661200MG Supplier TCI America Supplier No. D4661200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 544436-47-7
Molecular Formula C24H31Br2N
Molecular Weight (g/mol) 493.327
InChI Key NBJGUMLGJPJNLO-UHFFFAOYSA-N
PubChem CID 11191094
IUPAC Name 2,7-dibromo-9-dodecylcarbazole
SMILES CCCCCCCCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 493.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-dodecylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.