accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 544450-75-1

IUPAC Name: 4-chloro-N-(2-ethylhexyl)butanamide
Synonyms: 4-chloro-N-(2-ethylhexyl)butanamide
Molecular Weight (g/mol): 233.78
Molecular Formula: C12H24ClNO
InChi Key: OEPGTWXYVQAXOC-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNC(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results