Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-(2-ethylhexyl)butyramide, 97%, Thermo Scientific™

Catalog No. AAH59097MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59097MD 250 mg
AAH5909703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59097MD Supplier Thermo Scientific Chemicals Supplier No. H59097MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544450-75-1
Molecular Formula C12H24ClNO
Molecular Weight (g/mol) 233.78
MDL Number MFCD03379008
InChI Key OEPGTWXYVQAXOC-UHFFFAOYSA-N
Synonym 4-chloro-n-2-ethylhexyl butanamide, 4-chloro-n-2-ethylhexyl butyramide
PubChem CID 3516107
IUPAC Name 4-chloro-N-(2-ethylhexyl)butanamide
SMILES CCCCC(CC)CNC(=O)CCCCl

Specifications

Quantity 250 mg
Formula Weight 233.78
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(2-ethylhexyl)butyramide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.