accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5445-17-0

IUPAC Name: methyl 2-bromopropanoate
Synonyms: methyl 2-bromopropanoate
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: ACEONLNNWKIPTM-UHFFFAOYNA-N
SMILES: COC(=O)C(C)Br
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)