Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromopropionate 98.0+%, TCI America™
SDP

Catalog No. B146025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B146025G 25 g
B1460100G 100 g
B1460500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B146025G Supplier TCI America Supplier No. B146025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5445-17-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.002
MDL Number MFCD00000143
InChI Key ACEONLNNWKIPTM-UHFFFAOYSA-N
PubChem CID 95576
IUPAC Name methyl 2-bromopropanoate
SMILES CC(C(=O)OC)Br

Specifications

Color Yellow
Boiling Point 144°C
Quantity 25 g
UN Number 2920
Synonym methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester
Formula Weight 167.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromopropionate
EINECSNumber (2)-1179
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.